siRNA Details
Gene Symbol: Padi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019332.1 | NP_062205.1 | 10 | 1267 | ||
GenBank mRNA | AB010998.1 | - | 10 | 1267 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Padi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019332.1 | NP_062205.1 | 10 | 1267 | ||
GenBank mRNA | AB010998.1 | - | 10 | 1267 |