siRNA Details
| Gene Symbol: Rgs12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019339.1 | NP_062212.1 | 1 | 1916 | ||
| XM_006251351.2 | XP_006251413.1 | 2 | 2048 | |||
| XM_006251352.2 | XP_006251414.1 | 2 | 2052 | |||
| XM_006251353.2 | XP_006251415.1 | 2 | 2572 | |||
| XM_008770350.1 | XP_008768572.1 | 2 | 2585 | |||
| GenBank mRNA | U92280.1 | - | 1 | 1916 | ||

Rgs12
Genomic Map