siRNA Details
| Gene Symbol: Rgs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019340.1 | NP_062213.1 | 11 | 1789 | ||
| XM_006238260.2 | XP_006238322.1 | 18 | 2372 | |||
| XM_008763800.1 | XP_008762022.1 | 7 | 1175 | |||
| XM_008763801.1 | XP_008762023.1 | 2 | 692 | |||
| XM_008763802.1 | XP_008762024.1 | 2 | 2131 | |||
| GenBank mRNA | AB055153.1 | - | 11 | 1789 | ||

Rgs3
Genomic Map