siRNA Details
| Gene Symbol: Slc12a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019345.3 | NP_062218.3 | 14 | 1785 | ||
| XM_008772323.1 | XP_008770545.1 | 15 | 1902 | |||
| XM_008772324.1 | XP_008770546.1 | 15 | 1902 | |||
| XM_008772325.1 | XP_008770547.1 | 15 | 1899 | |||
| XM_008772326.1 | XP_008770548.1 | 14 | 1815 | |||
| XM_008772327.1 | XP_008770549.1 | 14 | 1818 | |||
| XM_008772328.1 | XP_008770550.1 | 14 | 1815 | |||
| GenBank mRNA | U10097.2 | - | 14 | 1785 | ||

Slc12a3
Genomic Map