siRNA Details
Gene Symbol: Sstr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019348.1 | NP_062221.1 | 2 | 410 | ||
GenBank mRNA | M93273.1 | - | 2 | 410 | ||
M96817.1 | - | 2 | 410 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
