siRNA Details
| Gene Symbol: Sstr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019348.1 | NP_062221.1 | 2 | 1395 | ||
| GenBank mRNA | M93273.1 | - | 2 | 1395 | ||
| M96817.1 | - | 2 | 1395 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sstr2| Gene Symbol: Sstr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019348.1 | NP_062221.1 | 2 | 1395 | ||
| GenBank mRNA | M93273.1 | - | 2 | 1395 | ||
| M96817.1 | - | 2 | 1395 | |||
Set Membership: |
|