siRNA Details
| Gene Symbol: Sstr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019348.1 | NP_062221.1 | 2 | 1153 | ||
| GenBank mRNA | M93273.1 | - | 2 | 1153 | ||
| M96817.1 | - | 2 | 1153 | |||
| X98234.1 | - | 1 | 180 | |||
More Information
Other Species Targeted
Set Membership: |
|

Sstr2
Genomic Map