siRNA Details
| Gene Symbol: Slk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019349.2 | NP_062222.2 | 9 | 2104 | ||
| XM_006231558.2 | XP_006231620.1 | 9 | 2142 | |||
| XM_008760447.1 | XP_008758669.1 | 10 | 3401 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slk| Gene Symbol: Slk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019349.2 | NP_062222.2 | 9 | 2104 | ||
| XM_006231558.2 | XP_006231620.1 | 9 | 2142 | |||
| XM_008760447.1 | XP_008758669.1 | 10 | 3401 | |||