siRNA Details
| Gene Symbol: Tpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019353.2 | NP_062226.2 | 14, 15 | 2513 | ||
| XM_008764607.1 | XP_008762829.1 | 13, 14 | 2471 | |||
| XM_008764608.1 | XP_008762830.1 | 14, 15 | 2507 | |||
| GenBank mRNA | M31655.1 | - | 10, 11 | 2039 | ||
| X17396.1 | - | 14, 15 | 2512 | |||

Tpo
Genomic Map