siRNA Details
| Gene Symbol: Ucp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019354.3 | NP_062227.2 | 3 | 454 | ||
| GenBank mRNA | AB005143.1 | - | 1 | 81 | ||
| AB006613.1 | - | 1 | 81 | |||
| AB010743.1 | - | 3 | 425 | |||
| AF039033.1 | - | 1 | 81 | |||
| BC062230.1 | - | 3 | 426 | 6920447 | ||
| FQ227097.1 | - | 3 | 454 | |||

Ucp2
Genomic Map