siRNA Details
| Gene Symbol: Scfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019364.1 | NP_062237.1 | 7 | 566 | ||
| XM_006240085.2 | XP_006240147.1 | 7 | 615 | |||
| GenBank mRNA | D79221.1 | - | 7 | 566 | ||
| U35364.1 | - | 7 | 586 | |||
| U57687.1 | - | 7 | 553 | |||

Scfd1
Genomic Map