siRNA Details
Gene Symbol: Ppt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019367.2 | NP_062240.1 | 3 | 345 | ||
XM_006256017.1 | XP_006256079.1 | 3 | 660 | |||
XM_006256018.2 | XP_006256080.1 | 3 | 1482 | |||
XM_006256019.2 | XP_006256081.1 | 3 | 1215 | |||
XM_006256020.2 | XP_006256082.1 | 3 | 1070 | |||
XM_006256022.2 | XP_006256084.1 | 3 | 1029 | |||
XM_006256023.2 | XP_006256085.1 | 3 | 503 | |||
XM_008772762.1 | XP_008770984.1 | 3 | 1969 | |||
GenBank mRNA | AF061971.1 | - | 3 | 277 | ||
AF067790.1 | - | 3 | 317 | |||
BC062023.1 | - | 3 | 307 | 5623515 | ||
CB736242.1 | - | 3 | 400 | |||
FQ234132.1 | - | 3 | 287 |
