siRNA Details
| Gene Symbol: Ppt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019367.2 | NP_062240.1 | 5 | 624 | ||
| XM_006256017.1 | XP_006256079.1 | 5 | 939 | |||
| XM_006256018.2 | XP_006256080.1 | 5 | 1761 | |||
| XM_006256019.2 | XP_006256081.1 | 5 | 1494 | |||
| XM_006256020.2 | XP_006256082.1 | 5 | 1349 | |||
| XM_006256022.2 | XP_006256084.1 | 5 | 1308 | |||
| XM_006256023.2 | XP_006256085.1 | 5 | 782 | |||
| XM_008772762.1 | XP_008770984.1 | 5 | 2248 | |||
| GenBank mRNA | AF061971.1 | - | 5 | 556 | ||
| BC062023.1 | - | 5 | 586 | 5623515 | ||
| FQ234132.1 | - | 5 | 566 | |||

Ppt2
Genomic Map