siRNA Details
| Gene Symbol: Uso1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019379.1 | NP_062252.1 | 12 | 1257 | ||
| XM_006250750.2 | XP_006250812.1 | 12 | 1278 | |||
| GenBank mRNA | FQ209746.1 | - | 4 | 446 | ||
| U14192.1 | - | 12 | 1172 | |||
| U15589.1 | - | 12 | 1257 | |||

Uso1
Genomic Map