siRNA Details
| Gene Symbol: Uso1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019379.1 | NP_062252.1 | 18 | 2164 | ||
| XM_006250750.2 | XP_006250812.1 | 19 | 2206 | |||
| GenBank mRNA | FQ209746.1 | - | 11 | 1374 | ||
| U14192.1 | - | 18 | 2079 | |||
| U15589.1 | - | 18 | 2164 | |||

Uso1
Genomic Map