siRNA Details
| Gene Symbol: Anxa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019905.1 | NP_063970.1 | 5 | 328 | ||
| GenBank mRNA | BC059136.1 | - | 5 | 347 | 6920034 | |
| FQ225322.1 | - | 5 | 344 | |||
| L13039.1 | - | 4 | 299 | |||
| S73559.1 | - | 5 | 330 | |||
| X66871.1 | - | 5 | 328 | |||

Anxa2
Genomic Map