siRNA Details
| Gene Symbol: Pclo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110797.1 | NP_001104267.1 | 13654 | |||
| NM_020098.1 | NP_064483.1 | 13654 | ||||
| GenBank mRNA | AF138789.2 | - | 13654 | |||
| AF227534.1 | - | 13654 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pclo| Gene Symbol: Pclo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110797.1 | NP_001104267.1 | 13654 | |||
| NM_020098.1 | NP_064483.1 | 13654 | ||||
| GenBank mRNA | AF138789.2 | - | 13654 | |||
| AF227534.1 | - | 13654 | ||||