siRNA Details
| Gene Symbol: Adam3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020302.1 | NP_064698.1 | 2 | 240 | ||
| GenBank mRNA | BC078839.1 | - | 2 | 146 | 7114315 | |
| Y07903.1 | - | 2 | 240 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adam3a| Gene Symbol: Adam3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020302.1 | NP_064698.1 | 2 | 240 | ||
| GenBank mRNA | BC078839.1 | - | 2 | 146 | 7114315 | |
| Y07903.1 | - | 2 | 240 | |||