siRNA Details
Gene Symbol: Adam3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020302.1 | NP_064698.1 | 19 | 2239 | ||
GenBank mRNA | BC078839.1 | - | 19 | 2145 | 7114315 | |
Y07903.1 | - | 19 | 2239 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Adam3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020302.1 | NP_064698.1 | 19 | 2239 | ||
GenBank mRNA | BC078839.1 | - | 19 | 2145 | 7114315 | |
Y07903.1 | - | 19 | 2239 |