siRNA Details
| Gene Symbol: Aadac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020538.1 | NP_065413.1 | 5 | 803 | ||
| GenBank mRNA | AF182426.1 | - | 5 | 803 | ||
| BC088143.1 | - | 5 | 804 | 7323135 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aadac| Gene Symbol: Aadac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020538.1 | NP_065413.1 | 5 | 803 | ||
| GenBank mRNA | AF182426.1 | - | 5 | 803 | ||
| BC088143.1 | - | 5 | 804 | 7323135 | ||