siRNA Details
| Gene Symbol: Uts2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020537.1 | NP_065412.1 | 1 | 681 | ||
| GenBank mRNA | AB012210.1 | - | 1 | 681 | ||
| AB029611.1 | - | 1 | 681 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uts2r| Gene Symbol: Uts2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020537.1 | NP_065412.1 | 1 | 681 | ||
| GenBank mRNA | AB012210.1 | - | 1 | 681 | ||
| AB029611.1 | - | 1 | 681 | |||
Set Membership: |
|