siRNA Details
| Gene Symbol: Glrx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032614.2 | NP_116003.2 | 7 | 750 | ||
| XM_006230497.2 | XP_006230559.1 | 7 | 1163 | |||
| GenBank mRNA | AF118651.1 | - | 7 | 584 | ||
| BC086381.1 | - | 7 | 750 | 7301390 | ||
| FQ216152.1 | - | 4 | 400 | |||

Glrx3
Genomic Map