siRNA Details
| Gene Symbol: Glrx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032614.2 | NP_116003.2 | 9, 10 | 877 | ||
| XM_006230497.2 | XP_006230559.1 | 9, 10 | 1290 | |||
| GenBank mRNA | AF118651.1 | - | 9, 10 | 711 | ||
| BC086381.1 | - | 9, 10 | 877 | 7301390 | ||
| FQ216152.1 | - | 6, 7 | 527 | |||

Glrx3
Genomic Map