siRNA Details
| Gene Symbol: Sec23a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105732.2 | NP_001099202.1 | 18, 19 | 2243 | ||
| XM_006240121.2 | XP_006240183.1 | 18, 19 | 2347 | |||
| XM_008764661.1 | XP_008762883.1 | 18, 19 | 2355 | |||
| XM_008764662.1 | XP_008762884.1 | 18, 19 | 2351 | |||
| XM_008764663.1 | XP_008762885.1 | 17, 18 | 2175 | |||
| GenBank mRNA | BC169006.1 | - | 18, 19 | 2243 | 7189145 | |

Sec23a
Genomic Map