siRNA Details
| Gene Symbol: Sec23a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105732.2 | NP_001099202.1 | 12 | 1463 | ||
| XM_006240121.2 | XP_006240183.1 | 12 | 1567 | |||
| XM_008764661.1 | XP_008762883.1 | 12 | 1575 | |||
| XM_008764662.1 | XP_008762884.1 | 12 | 1571 | |||
| XM_008764663.1 | XP_008762885.1 | 11 | 1395 | |||
| GenBank mRNA | BC169006.1 | - | 12 | 1463 | 7189145 | |

Sec23a
Genomic Map