siRNA Details
Gene Symbol: Txnrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031614.2 | NP_113802.2 | 7 | 758 | ||
GenBank mRNA | AF108213.1 | - | 6 | 600 | ||
AF220760.1 | - | 6 | 654 | |||
BC085726.1 | - | 855 | 7112649 | |||
FQ225805.1 | - | 4 | 403 | |||
U63923.2 | - | 6 | 649 |
