siRNA Details
Gene Symbol: Txnrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031614.2 | NP_113802.2 | 8 | 907 | ||
GenBank mRNA | AF108213.1 | - | 7 | 749 | ||
AF220760.1 | - | 7 | 803 | |||
BC085726.1 | - | 1004 | 7112649 | |||
FQ225805.1 | - | 5 | 552 | |||
U63923.2 | - | 7 | 798 |
