siRNA Details
| Gene Symbol: Zfp148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031615.1 | NP_113803.1 | 4 | 700 | ||
| XM_006248435.2 | XP_006248497.1 | 1 | 330 | |||
| GenBank mRNA | U30381.1 | - | 4 | 700 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp148| Gene Symbol: Zfp148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031615.1 | NP_113803.1 | 4 | 700 | ||
| XM_006248435.2 | XP_006248497.1 | 1 | 330 | |||
| GenBank mRNA | U30381.1 | - | 4 | 700 | ||