siRNA Details
| Gene Symbol: Nr4a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031628.1 | NP_113816.1 | 5 | 1938 | ||
| XM_008763702.1 | XP_008761924.1 | 6 | 3732 | |||
| XM_008763703.1 | XP_008761925.1 | 5 | 3660 | |||
| GenBank mRNA | D38530.1 | - | 5 | 1938 | ||
| DQ268830.1 | - | 5 | 1938 | |||
| X86003.1 | - | 3 | 1725 | |||

Nr4a3
Genomic Map