siRNA Details
Gene Symbol: Ddx25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031630.2 | NP_113818.2 | 11 | 1365 | ||
GenBank mRNA | AF142629.2 | - | 11 | 1365 | ||
BC078791.1 | - | 11 | 1347 | 7112991 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ddx25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031630.2 | NP_113818.2 | 11 | 1365 | ||
GenBank mRNA | AF142629.2 | - | 11 | 1365 | ||
BC078791.1 | - | 11 | 1347 | 7112991 |