siRNA Details
| Gene Symbol: Ccnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_171992.4 | NP_741989.3 | 2 | 462 | ||
| XM_008760168.1 | XP_008758390.1 | 2 | 637 | |||
| GenBank mRNA | AB256048.1 | - | 2 | 515 | ||
| AM258963.2 | - | 2 | 515 | |||
| CA510842.1 | - | 2 | 463 | |||
| D14014.1 | - | 2 | 462 | |||
| FQ231654.1 | - | 2 | 475 | |||
| X75207.1 | - | 2 | 489 | |||

Ccnd1
Genomic Map