siRNA Details
| Gene Symbol: Plk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022187.1 | NP_071523.1 | 10, 11 | 1368 | ||
| GenBank mRNA | AF136584.1 | - | 10, 11 | 1164 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plk3| Gene Symbol: Plk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022187.1 | NP_071523.1 | 10, 11 | 1368 | ||
| GenBank mRNA | AF136584.1 | - | 10, 11 | 1164 | ||