siRNA Details
| Gene Symbol: Hgfac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053320.1 | NP_445772.1 | 2 | 151 | ||
| XM_008770352.1 | XP_008768574.1 | 2 | 254 | |||
| GenBank mRNA | BC089871.1 | - | 2 | 151 | 7324508 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hgfac| Gene Symbol: Hgfac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053320.1 | NP_445772.1 | 2 | 151 | ||
| XM_008770352.1 | XP_008768574.1 | 2 | 254 | |||
| GenBank mRNA | BC089871.1 | - | 2 | 151 | 7324508 | |