siRNA Details
| Gene Symbol: Dlg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031639.1 | NP_113827.1 | 19 | 2635 | ||
| XM_006257080.2 | XP_006257142.1 | 19 | 2962 | |||
| XM_006257081.2 | XP_006257143.1 | 18 | 2907 | |||
| XM_006257082.2 | XP_006257144.1 | 18 | 2870 | |||
| XM_006257083.2 | XP_006257145.1 | 17 | 2814 | |||
| XM_006257084.2 | XP_006257146.1 | 12 | 1362 | |||
| XM_006257085.2 | XP_006257147.1 | 10 | 1753 | |||
| XM_008773267.1 | XP_008771489.1 | 18 | 2922 | |||
| XM_008773268.1 | XP_008771490.1 | 12 | 1271 | |||
| GenBank mRNA | U50147.1 | - | 19 | 2635 | ||

Dlg3
Genomic Map