siRNA Details
| Gene Symbol: Hpgds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031644.2 | NP_113832.1 | 5, 6 | 798 | ||
| GenBank mRNA | AF021882.1 | - | 4, 5 | 326 | ||
| BC087590.1 | - | 5, 6 | 798 | 7307804 | ||
| D82071.1 | - | 5, 6 | 629 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hpgds| Gene Symbol: Hpgds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031644.2 | NP_113832.1 | 5, 6 | 798 | ||
| GenBank mRNA | AF021882.1 | - | 4, 5 | 326 | ||
| BC087590.1 | - | 5, 6 | 798 | 7307804 | ||
| D82071.1 | - | 5, 6 | 629 | |||