siRNA Details
| Gene Symbol: Nme6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191884.1 | NP_001178813.1 | 6, 7 | 617 | ||
| XM_006243847.2 | XP_006243909.1 | 6, 7 | 626 | |||
| XM_006243848.2 | XP_006243910.1 | 6, 7 | 552 | |||
| XM_006243849.1 | XP_006243911.1 | 6, 7 | 1070 | |||
| XM_006243850.2 | XP_006243912.1 | 6, 7 | 591 | |||
| XM_006243851.1 | XP_006243913.1 | 6, 7 | 655 | |||
| GenBank mRNA | AF051943.1 | - | 4, 5 | 359 | ||
| FQ225386.1 | - | 6, 7 | 796 | |||

Nme6
Genomic Map