siRNA Details
| Gene Symbol: Nme6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191884.1 | NP_001178813.1 | 6 | 540 | ||
| XM_006243847.2 | XP_006243909.1 | 6 | 549 | |||
| XM_006243848.2 | XP_006243910.1 | 6 | 475 | |||
| XM_006243849.1 | XP_006243911.1 | 6 | 993 | |||
| XM_006243850.2 | XP_006243912.1 | 6 | 514 | |||
| XM_006243851.1 | XP_006243913.1 | 6 | 578 | |||
| GenBank mRNA | AF051943.1 | - | 4 | 282 | ||
| FQ225386.1 | - | 6 | 719 | |||

Nme6
Genomic Map