siRNA Details
| Gene Symbol: Nme6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191884.1 | NP_001178813.1 | 7 | 663 | ||
| XM_006243847.2 | XP_006243909.1 | 7 | 672 | |||
| XM_006243848.2 | XP_006243910.1 | 7 | 598 | |||
| XM_006243849.1 | XP_006243911.1 | 7 | 1116 | |||
| XM_006243850.2 | XP_006243912.1 | 7 | 637 | |||
| XM_006243851.1 | XP_006243913.1 | 7 | 701 | |||
| GenBank mRNA | AF051943.1 | - | 5 | 405 | ||
| FQ225386.1 | - | 7 | 842 | |||

Nme6
Genomic Map