siRNA Details
Gene Symbol: Sfmbt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031647.2 | NP_113835.2 | 14, 15 | 1682 | ||
XM_006252656.2 | XP_006252718.1 | 14, 15 | 1968 | |||
XM_006252657.2 | XP_006252719.1 | 15, 16 | 2028 | |||
XM_006252658.1 | XP_006252720.1 | 14, 15 | 3940 | |||
XM_008771003.1 | XP_008769225.1 | 16, 17 | 2949 | |||
XM_008771004.1 | XP_008769226.1 | 15, 16 | 2889 | |||
XM_008771005.1 | XP_008769227.1 | 17, 18 | 2258 | |||
XM_008771006.1 | XP_008769228.1 | 16, 17 | 2122 | |||
GenBank mRNA | AB032164.1 | - | 14, 15 | 1676 | ||
BC081770.1 | - | 14, 15 | 1682 | 7132495 |
