siRNA Details
| Gene Symbol: Slc30a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022853.2 | NP_074044.1 | 2 | 950 | ||
| GenBank mRNA | U17133.1 | - | 2 | 951 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc30a1| Gene Symbol: Slc30a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022853.2 | NP_074044.1 | 2 | 950 | ||
| GenBank mRNA | U17133.1 | - | 2 | 951 | ||