siRNA Details
Gene Symbol: Slc13a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031651.1 | NP_113839.1 | 8, 9 | 952 | ||
GenBank mRNA | L19102.1 | - | 8, 9 | 952 | ||
U08031.1 | - | 936 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc13a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031651.1 | NP_113839.1 | 8, 9 | 952 | ||
GenBank mRNA | L19102.1 | - | 8, 9 | 952 | ||
U08031.1 | - | 936 |