siRNA Details
| Gene Symbol: P3h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021581.1 | NP_067592.1 | 7 | 1097 | ||
| XM_006247414.2 | XP_006247476.1 | 7 | 1401 | |||
| GenBank mRNA | FQ227296.1 | - | 7 | 1203 | ||
| X65454.1 | - | 7 | 1097 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

P3h4| Gene Symbol: P3h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021581.1 | NP_067592.1 | 7 | 1097 | ||
| XM_006247414.2 | XP_006247476.1 | 7 | 1401 | |||
| GenBank mRNA | FQ227296.1 | - | 7 | 1203 | ||
| X65454.1 | - | 7 | 1097 | |||