siRNA Details
| Gene Symbol: Ntrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021589.1 | NP_067600.1 | 5, 6 | 632 | ||
| XM_006232746.2 | XP_006232808.1 | 5, 6 | 623 | |||
| GenBank mRNA | AF174586.1 | - | 2, 3 | 226 | ||
| AF174587.1 | - | 1, 2 | 157 | |||
| M85214.1 | - | 5, 6 | 632 | |||

Ntrk1
Genomic Map