siRNA Details
| Gene Symbol: Kmo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021593.1 | NP_067604.1 | 4, 5 | 347 | ||
| XM_006250325.2 | XP_006250387.1 | 4, 5 | 512 | |||
| XM_006250327.2 | XP_006250389.1 | 3, 4 | 417 | |||
| XM_008769794.1 | XP_008768016.1 | 2, 3 | 417 | |||
| GenBank mRNA | AF056031.1 | - | 4, 5 | 347 | ||
| BC088142.1 | - | 4, 5 | 328 | 7322842 | ||
| FQ210049.1 | - | 4, 5 | 345 | |||

Kmo
Genomic Map