siRNA Details
| Gene Symbol: Nucb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021663.2 | NP_067695.1 | 13, 14 | 1462 | ||
| XM_006230095.2 | XP_006230157.1 | 12, 13 | 1370 | |||
| GenBank mRNA | AF238223.1 | - | 11 | 1249 | ||
| AF250142.1 | - | 12, 13 | 1349 | |||
| BC061778.1 | - | 13, 14 | 1462 | 5621784 | ||

Nucb2
Genomic Map