siRNA Details
| Gene Symbol: Dnase2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021664.1 | NP_067696.1 | 1 | 232 | ||
| XM_006233464.2 | XP_006233526.1 | 2 | 309 | |||
| XM_006233465.2 | XP_006233527.1 | 2 | 336 | |||
| XM_006233466.2 | XP_006233528.1 | 2 | 339 | |||
| XM_006233468.2 | XP_006233530.1 | 2 | 720 | |||
| XM_008761521.1 | XP_008759743.1 | 2 | 442 | |||
| GenBank mRNA | AF178974.1 | - | 1 | 232 | ||

Dnase2b
Genomic Map