siRNA Details
| Gene Symbol: Dnase2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021664.1 | NP_067696.1 | 5 | 848 | ||
| XM_006233464.2 | XP_006233526.1 | 6 | 925 | |||
| XM_006233465.2 | XP_006233527.1 | 6 | 952 | |||
| XM_006233466.2 | XP_006233528.1 | 6 | 955 | |||
| XM_006233468.2 | XP_006233530.1 | 6 | 1336 | |||
| XM_008761521.1 | XP_008759743.1 | 6 | 1058 | |||
| GenBank mRNA | AF178974.1 | - | 5 | 848 | ||

Dnase2b
Genomic Map