siRNA Details
| Gene Symbol: Dnase2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021664.1 | NP_067696.1 | 6 | 1106 | ||
| XM_006233464.2 | XP_006233526.1 | 7 | 1183 | |||
| XM_006233465.2 | XP_006233527.1 | 7 | 1210 | |||
| XM_006233466.2 | XP_006233528.1 | 7 | 1213 | |||
| XM_006233468.2 | XP_006233530.1 | 7 | 1594 | |||
| XM_008761521.1 | XP_008759743.1 | 7 | 1316 | |||
| GenBank mRNA | AF178974.1 | - | 6 | 1106 | ||

Dnase2b
Genomic Map