siRNA Details
| Gene Symbol: Epb41l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021681.2 | NP_067713.2 | 24 | 4736 | ||
| NM_172090.3 | NP_742087.1 | 21 | 2703 | |||
| XM_008762381.1 | XP_008760603.1 | 24 | 4736 | |||
| GenBank mRNA | AB019256.1 | - | 21 | 2701 | ||
| AB019257.1 | - | 24 | 4736 | |||

Epb41l1
Genomic Map