siRNA Details
| Gene Symbol: Rapgef3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021690.1 | NP_067722.1 | 6 | 784 | ||
| GenBank mRNA | U78167.1 | - | 6 | 784 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rapgef3| Gene Symbol: Rapgef3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021690.1 | NP_067722.1 | 6 | 784 | ||
| GenBank mRNA | U78167.1 | - | 6 | 784 | ||